Please login to your online account to display your discounted pricing

anti-ERAS, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223727

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223727

  • / Each
 

Description & Specifications

Specifications

Antigen ERAS
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. Q7Z444
Host Species Rabbit
Immunogen Within the range of amino acids 178-210 of human ERAS protein were used as the immunogen.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3266
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
Target Species Human
Target Species Rat