Please login to your online account to display your discounted pricing

anti-ERAS, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 178-210 of human ERAS protein were used as the immunogen.
Antigen ERAS
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223727

 
novus biologicals canada
NBP223727
Unlabeled 0.1mL Each for $257.41

NBP223727SS

 
novus biologicals canada
NBP223727SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 178-210 of human ERAS protein were used as the immunogen.
Antigen ERAS
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
Gene Accession No. Q7Z444
Gene ID (Entrez) 3266
Formulation 100μL of crude rabbit sera with 50% glycerol