Please login to your online account to display your discounted pricing

anti-EPS15, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11074689SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11074689T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P42566
Monoclonal or Polyclonal Polyclonal
Concentration 0.75mg/mL
Formulation PBS with 0.1% BSA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Format Affinity Purified
Conjugate Unlabeled
Gene ID (Entrez) 2060
Immunogen Synthetic peptide made to the C-terminal portion of human EPS15 (within residues 840-896). [Swiss-Prot No. P42566]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.025mL
Antigen EPS15
Primary or Secondary Primary
Host Species Rabbit
Regulatory Status RUO