Please login to your online account to display your discounted pricing

anti-EPS15, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11074689

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11074689

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.1% BSA
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P42566
Conjugate Unlabeled
Concentration 0.75mg/mL
Gene ID (Entrez) 2060
Format Affinity Purified
Immunogen Synthetic peptide made to the C-terminal portion of human EPS15 (within residues 840-896). [Swiss-Prot No. P42566]
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL
Antigen EPS15
Regulatory Status RUO