Please login to your online account to display your discounted pricing

anti-EPS15, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to the C-terminal portion of human EPS15 (within residues 840-896). [Swiss-Prot No. P42566]
Antigen EPS15
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11074689T

 
novus biologicals canada
NB11074689SS
Unlabeled 0.025mL Each for $139.00

NB11074689

 
novus biologicals canada
NB11074689
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to the C-terminal portion of human EPS15 (within residues 840-896). [Swiss-Prot No. P42566]
Antigen EPS15
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P42566
Gene ID (Entrez) 2060
Formulation PBS with 0.1% BSA
Concentration 0.75mg/mL