Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227107SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227107SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 4072
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat.
Immunogen A portion of amino acids 250-350 of human EpCAM/CD326 was used as the immunogen.
Antigen EpCAM/TROP1
Regulatory Status RUO
Format Purified
Primary or Secondary Primary
Quantity 0.025mg
Host Species Rabbit