Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227107

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227107

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4072
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat.
Regulatory Status RUO
Antigen EpCAM/TROP1
Concentration 0.5mg/mL
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Format Purified
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Immunogen A portion of amino acids 250-350 of human EpCAM/CD326 was used as the immunogen.
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg