Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227361SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227361SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Primary or Secondary Primary
Immunogen A portion of amino acids 130-180 of human EpCAM was used as the immunogen for this antibody.
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat. Per, there are two isoforms of EpCAM: 314 amino acids (∽33 kD) and 342 amino acids (∽38 kD). IMG-6753A detects both isoforms while detects the 342 amino acid form.
Gene Accession No. O55159
Gene ID (Entrez) 4072
Host Species Rabbit
Regulatory Status RUO
Format Purified
Quantity 0.025mg
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen EpCAM/TROP1
Cross Reactivity Human