Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227361

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227361

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O55159
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4072
Conjugate Unlabeled
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Primary or Secondary Primary
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat. Per, there are two isoforms of EpCAM: 314 amino acids (∽33 kD) and 342 amino acids (∽38 kD). IMG-6753A detects both isoforms while detects the 342 amino acid form.
Cross Reactivity Human
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 130-180 of human EpCAM was used as the immunogen for this antibody.
Host Species Rabbit
Antigen EpCAM/TROP1
Quantity 0.1mg