Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human EpCAM was used as the immunogen for this antibody.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227361SS

 
novus biologicals canada
NBP227361SS
Unlabeled 0.025mg Each for $139.00

NBP227361

 
novus biologicals canada
NBP227361
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human EpCAM was used as the immunogen for this antibody.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat. Per, there are two isoforms of EpCAM: 314 amino acids (∽33 kD) and 342 amino acids (∽38 kD). IMG-6753A detects both isoforms while detects the 342 amino acid form.
Gene Accession No. O55159
Gene ID (Entrez) 4072
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL