Please login to your online account to display your discounted pricing

anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-350 of human EpCAM/CD326 was used as the immunogen.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227107

 
novus biologicals canada
NBP227107
Unlabeled 0.1mg Each for $257.41

NBP227107SS

 
novus biologicals canada
NBP227107SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-350 of human EpCAM/CD326 was used as the immunogen.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat.
Gene ID (Entrez) 4072
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL