Please login to your online account to display your discounted pricing

anti-Enolase 2/Neuron-specific Enolase, Polyclonal, Novus Biologicals™

$219.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Neuron specific enolase (gamma-gamma isoenzyme) isolated from bovine brain.
Antigen Enolase 2/Neuron-specific Enolase
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12083460M

 
novus biologicals canada
NB12083460ML
Unlabeled 6mL Each for $309.05

NB12083405M

 
novus biologicals canada
NB12083405ML
Unlabeled 0.5mL Each for $219.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Neuron specific enolase (gamma-gamma isoenzyme) isolated from bovine brain.
Antigen Enolase 2/Neuron-specific Enolase
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity This antibody reacts with human isoenzymes of neuron specific enolase containing gamma subunits. It gives a specific staining in neurons of the brain, islets of Langerhans in pancreas, and in peripheral nerves of the skin. No staining with other tissue types has been seen. Not yet tested in other species.
Gene ID (Entrez) 2026