Please login to your online account to display your discounted pricing

anti-Enolase 2/Neuron-specific Enolase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12083460ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12083460M

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Primary or Secondary Primary
Gene ID (Entrez) 2026
Immunogen Neuron specific enolase (gamma-gamma isoenzyme) isolated from bovine brain.
Antigen Enolase 2/Neuron-specific Enolase
Host Species Rabbit
Test Specificity This antibody reacts with human isoenzymes of neuron specific enolase containing gamma subunits. It gives a specific staining in neurons of the brain, islets of Langerhans in pancreas, and in peripheral nerves of the skin. No staining with other tissue types has been seen. Not yet tested in other species.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Format Antiserum
Quantity 6mL
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO