missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Endophilin-A1 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT22
Description
This Anti-Endophilin-A1 Antibody is validated for use in western blotting for the detection of Endophilin-A1.Specifications
Endophilin-A1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide and 50% glycerol. | |
Rabbit | |
Affinity Purified | |
Primary | |
Mouse, Rat | |
IgG |
Western Blot | |
Unconjugated | |
SH3D2A; CNSA2; EEN-B11; SH3P4 | |
KLH-conjugated linear peptide corresponding to mouse Endophilin-A1. | |
100 μL | |
NP_062408 | |
2-8°C for 12 months |
For Research Use Only