Please login to your online account to display your discounted pricing

anti-Encephalopsin/OPN3 Polyclonal, Thermo Scientific™

Encephalopsin/OPN3 Polyclonal Ab PA5-34038 from Thermo Scientific™ specifically detects Encephalopsin/OPN3 in Hu samples, and it is validated for IHC(P)

$506.05 - $506.05

Specifications

Antigen Encephalopsin/OPN3
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human OPN3/Encephalopsin.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA534038

 
pierce biotechnology inc
PA534038
Unlabeled 50μg Each for $506.05

PIPA534039

 
pierce biotechnology inc
PA534039
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen Encephalopsin/OPN3
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human OPN3/Encephalopsin.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

Encephalopsin/OPN3 Polyclonal Ab PA5-34038 from Thermo Scientific™ specifically detects Encephalopsin/OPN3 in Hu samples, and it is validated for IHC(P)

Potential odorant receptor.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from C-terminus of human OPN3/Encephalopsin.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: OPN3, ERO, Encephalopsin, NMO-1, Opsin 3, Opsin-3, ECPN, Panopsin
Uniprot Accession: Q9H1Y3
Gene Symbol: OPN3
Entrez GeneID: 23596
PA5-34046 detects OR10R2 in Human samples. PA5-34046 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-34046 The immunogen is Synthetic 14 amino acid peptide from 2nd extracellular domain of human OR10R2..