Please login to your online account to display your discounted pricing

anti-EMAP-II/AIMP1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 720-800 from human AIMP1 was used as the immunogen for this antibody.
Antigen EMAP-II/AIMP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227206SS

 
novus biologicals canada
NBP227206SS
Unlabeled 0.025mg Each for $109.00

NBP227206

 
novus biologicals canada
NBP227206
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 720-800 from human AIMP1 was used as the immunogen for this antibody.
Antigen EMAP-II/AIMP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, orangutan, panda, cow, guinea pig, new world monkey, dog, Chinese hamster, horse, naked mole rat, rhesus monkey, opossum, rabbit, pig, cat, and white-cheeked gibbon, 94% in chimpanzee, frog, rat, platypus, wild turkey, elephant, chicken, and Carolina anole, and 83% homologous in zebrafish, cichlid fish, and catfish.
Gene ID (Entrez) 9255
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL