Please login to your online account to display your discounted pricing

anti-EMAP-II/AIMP1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 260-310 from human AIMP1 was used as the immunogen for this antibody.
Antigen EMAP-II/AIMP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227080SS

 
novus biologicals canada
NBP227080SS
Unlabeled 0.025mg Each for $109.00

NBP227080

 
novus biologicals canada
NBP227080
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 260-310 from human AIMP1 was used as the immunogen for this antibody.
Antigen EMAP-II/AIMP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, orangutan, cow, guinea pig, Chinese hamster, horse, naked mole rat, rhesus monkey, pig, african bush elephant, white-cheeked gibbon, and chimpanzee, 94% in panda, rabbit, platypus, wild turkey, and chicken, 89% in new world monkey, dog, opossum, cat, frog, zebrafish, cichlid fish, and catfish, 83% homologous in Carolina anole.
Gene ID (Entrez) 9255
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL