missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
elongation protein 4 homolog (S. cerevisiae) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ELP4 protein.
Supplier: Abnova Corporation H00026610B01
Description
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq
Sequence: MAAVATCGSVAASTGSAVATASKSNVTSFQRRGPRASVTNDSGPRLVSIAGTRPSVRNGQLLVSTGLPALDQLLGGGLAVGTVLLIEEDKYNIYSPLLFKYFLAEGIVNGHTLLVASAKEDPANILQELPAPLLDDKCKKEFDEDVYNHKTPESNIKMKIAWRYQLLPKMEIGPVSSSRFGHYYDASKRMPQELIEASNWHGFFLPEKISSTLKVEPCSLTPGYTKLLQFIQNIIYEEGFDGSNPQKKQRNILRIGIQNLGSPLWGDDICCAENGGNSHSLTKFLYVLRGLLRTSLSACIITMPTHLIQNKAIIARVTTLSDVVVGLESFIGSERETNPLYKDYHGLIHIRQIPRLNNLICDESDVKDLAFKLKRKLFTIEAGVQWHDLGSRRPRLLGSGGSPASASLVAGITGAHHHAQLIFVFLVEMGFHHVGQAGLELLTSGDSSASASQSAGIAGMSYRARPRALYFKENKSKVGARQLLETREEHLSSRLLILTQAERLCMGRRFFTAFHIFNELPCKGDCICLQTCQTQSpecifications
elongation protein 4 homolog (S. cerevisiae) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ELP4 protein. | |
ELP4 | |
C11orf19/FLJ20498/PAX6NEB/PAXNEB/dJ68P15A.1 | |
Mouse | |
50 μL | |
Kinases | |
Primary | |
Human | |
Serum |
Immunofluorescence,Western Blot | |
Unconjugated | |
No additive | |
BC012514.1 | |
ELP4 | |
ELP4 (AAH12514.1, 1 a.a. ∼ 535 a.a) full-length human protein. | |
RUO | |
Yes | |
26610 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |