missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EphB1 Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA112410

Catalog No. 01-674-139



Description

Description

A suggested positive control for this product is human brain lysate.

ephB1, previously known as Elk(eph-like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. ephB family proteins bind ephrins of the B class, ligands that are also transmembrane spanning proteins capable of transmitting signals. ephB1 is expressed predomitly in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, ephB1 signals regulation of cell attachment and cellcell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.
Specifications

Specifications

EphB1
Polyclonal
PBS with 0.08% sodium azide
P54762
ELK, EPHT2, FLJ37986, Hek6, NET
Sheep
IgG
250 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2047
Immunoprecipitation
Unconjugated
EPHB1
Liquid
EPHB1
Recombit protein corresponding to residues 883-984 of C terminus of human ephB1.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents