Please login to your online account to display your discounted pricing

anti-ELK Rec. Tyrosine Kinase Polyclonal, Thermo Scientific™

ELK Rec. Tyrosine Kinase Polyclonal Ab PA1-12409 from Thermo Scientific™ specifically detects ELK Rec. Tyrosine Kinase in Hu samples, and it is validated for IP

$448.05 - $448.05

Specifications

Antigen ELK Rec. Tyrosine Kinase
Cross Reactivity Human
Format Purified
Host Species Sheep
Immunogen Recombinant protein corresponding to residues 16-351 of N terminus of human ephB1.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-138

 
pierce biotechnology inc
PA112409
Unlabeled 250μg Each for $448.05

01-674-139

 
pierce biotechnology inc
PA112410
Unlabeled 250μg Each for $448.05

01-674-140

 
pierce biotechnology inc
PA112411
Unlabeled 250μg Each for $448.05
Description & Specifications

Specifications

Antigen ELK Rec. Tyrosine Kinase
Cross Reactivity Human
Format Purified
Host Species Sheep
Immunogen Recombinant protein corresponding to residues 16-351 of N terminus of human ephB1.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

ELK Rec. Tyrosine Kinase Polyclonal Ab PA1-12409 from Thermo Scientific™ specifically detects ELK Rec. Tyrosine Kinase in Hu samples, and it is validated for IP

ephB1, previously known as Elk(eph-like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. ephB family proteins bind ephrins of the B class, ligands that are also transmembrane spanning proteins capable of transmitting signals. ephB1 is expressed predominantly in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, ephB1 signals regulation of cell attachment and cellcell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.
Host Species: Sheep
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant protein corresponding to residues 16-351 of N terminus of human ephB1.
Formulation: 250μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: ELK, EPHT2, FLJ37986, Hek6, NET
Uniprot Accession: P54762
Gene Symbol: EPHB1
Entrez GeneID: 2047
PA1-12409 detects ELK Rec. Tyrosine Kinase in Human samples. PA1-12409 has been successfully used in Immunoprecipitation procedures. PA1-12409 The immunogen is Recombinant protein corresponding to residues 16-351 of N terminus of human ephB1..