missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-EIF3D, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human eIF3D was used as the immunogen.
Antigen EIF3D
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224468 Novus Biologicals Canada
NBP224468
0.1mg Each for $255.80
NBP224468SS Novus Biologicals Canada
NBP224468SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
EIF3D
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, opossum, mouse, cattle, rat and chimpanzee.
8664
0.5mg/mL
A portion of amino acids 500-550 of human eIF3D was used as the immunogen.
Polyclonal
Primary
Western Blot
O15371
100μg in 200μL PBS containing 0.05% BSA