Please login to your online account to display your discounted pricing

anti-EIF3B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224571SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224571SS

  • / Each
 

Description & Specifications

Specifications

Antigen EIF3B
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P55884
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human eIF3B was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8662
Test Specificity Reactive in human and mouse. Predicted to cross react with primate samples by homology.
Target Species Human
Target Species Murine