Please login to your online account to display your discounted pricing

anti-EIF3B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224571

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224571

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8662
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Regulatory Status RUO
Test Specificity Reactive in human and mouse. Predicted to cross react with primate samples by homology.
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P55884
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Antigen EIF3B
Primary or Secondary Primary
Immunogen A portion of amino acids 1-50 of human eIF3B was used as the immunogen for the antibody.