Please login to your online account to display your discounted pricing

anti-EIF3B, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human eIF3B was used as the immunogen for the antibody.
Antigen EIF3B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224571SS

 
novus biologicals canada
NBP224571SS
Unlabeled 0.025mg Each for $109.00

NBP224571

 
novus biologicals canada
NBP224571
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human eIF3B was used as the immunogen for the antibody.
Antigen EIF3B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Reactive in human and mouse. Predicted to cross react with primate samples by homology.
Gene Accession No. P55884
Gene ID (Entrez) 8662
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL