Please login to your online account to display your discounted pricing

anti-EGLN2/PHD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100310SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100310SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_444274.1
Gene ID (Entrez) 112398
Conjugate Unlabeled
Primary or Secondary Primary
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen EGLN2/PHD1
Host Species Rabbit
Regulatory Status RUO
Immunogen The epitope maps to a region between residue 350 and the C-terminus (residue 407) of human PHD1/HIF Prolyl Hydroxylase 1 using the numbering given in entry NP_444274.1 (GeneID 112398).