Please login to your online account to display your discounted pricing

anti-EGLN2/PHD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100310

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100310

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen EGLN2/PHD1
Conjugate Unlabeled
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Gene Accession No. NP_444274.1
Gene ID (Entrez) 112398
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Format Affinity Purified
Immunogen The epitope maps to a region between residue 350 and the C-terminus (residue 407) of human PHD1/HIF Prolyl Hydroxylase 1 using the numbering given in entry NP_444274.1 (GeneID 112398).
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit