Please login to your online account to display your discounted pricing

anti-EGLN2/PHD1, Polyclonal, Novus Biologicals™

$160.00 - $362.00

Specifications

Host Species Rabbit
Immunogen The epitope maps to a region between residue 350 and the C-terminus (residue 407) of human PHD1/HIF Prolyl Hydroxylase 1 using the numbering given in entry NP_444274.1 (GeneID 112398).
Antigen EGLN2/PHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100310SS

 
novus biologicals canada
NB100310SS
Unlabeled 0.025mL Each for $160.00

NB100310

 
novus biologicals canada
NB100310
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope maps to a region between residue 350 and the C-terminus (residue 407) of human PHD1/HIF Prolyl Hydroxylase 1 using the numbering given in entry NP_444274.1 (GeneID 112398).
Antigen EGLN2/PHD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Gene Accession No. NP_444274.1
Gene ID (Entrez) 112398
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL