Please login to your online account to display your discounted pricing

anti-EGLN1/PHD2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100137

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100137

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Antigen EGLN1/PHD2
Primary or Secondary Primary
Gene ID (Entrez) 54583
Test Specificity Human. This antibody does not appear to work in Mouse. Results have been mixed in Rat with success in western analysis and immunofluorescence on Rat endothelial cells and negative results with PC12 cells. Expected reactivity with Rabbit, Orangutan, Rhesus Monkey, Gorilla based on 100% sequence identity.
Gene Accession No. Q9GZT9
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Format Affinity Purified
Immunogen The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583).
Quantity 0.1mL
Regulatory Status RUO
Host Species Rabbit
Cross Reactivity Human