Please login to your online account to display your discounted pricing

anti-EGLN1/PHD2, Polyclonal, Novus Biologicals™

$160.00 - $362.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583).
Antigen EGLN1/PHD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100137

 
novus biologicals canada
NB100137
Unlabeled 0.1mL Each for $362.00

NB100137SS

 
novus biologicals canada
NB100137SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583).
Antigen EGLN1/PHD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Human. This antibody does not appear to work in Mouse. Results have been mixed in Rat with success in western analysis and immunofluorescence on Rat endothelial cells and negative results with PC12 cells. Expected reactivity with Rabbit, Orangutan, Rhesus Monkey, Gorilla based on 100% sequence identity.
Gene Accession No. Q9GZT9
Gene ID (Entrez) 54583
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL