Please login to your online account to display your discounted pricing

anti-EGLN1/PHD2, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400). [UniProt No. Q91YE3]
Antigen EGLN1/PHD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002219SS

 
novus biologicals canada
NB1002219SS
Unlabeled 0.025mL Each for $144.63

NB1002219

 
novus biologicals canada
NB1002219
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400). [UniProt No. Q91YE3]
Antigen EGLN1/PHD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human and Mouse. Predicted to react with rat based on 100% sequence homology.
Gene ID (Entrez) 54583
Formulation PBS
Concentration 1mg/mL