Please login to your online account to display your discounted pricing

anti-EGLN1/PHD2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100138SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100138SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 54583
Concentration 1mg/mL
Gene Accession No. Q9GZT9
Immunogen The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583).
Applications Electron Microscopy
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Test Specificity Human. Does not appear to work in mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Format Affinity Purified
Regulatory Status RUO
Quantity 0.025mL
Antigen EGLN1/PHD2
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal