Please login to your online account to display your discounted pricing

anti-EGF, Polyclonal, Novus Biologicals™

$397.00 - $397.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the C Terminus Region of the target protein.
Antigen EGF
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

26-610-002

 
novus biologicals canada
26610002
Unlabeled 0.05mg Each for $397.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the C Terminus Region of the target protein.
Antigen EGF
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Dot Blot
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity This product is specific for Human EGF.
Gene Accession No. P01133
Gene ID (Entrez) 1950
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0