Please login to your online account to display your discounted pricing

anti-EDIL3/DEL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP128632SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP128632SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O43854
Quantity 0.025mL
Concentration 1.62mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Gene ID (Entrez) 10085
Host Species Rabbit
Antigen EDIL3/DEL1
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot No. O43854]
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary