Please login to your online account to display your discounted pricing

anti-EDIL3/DEL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP128632

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP128632

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. O43854
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 10085
Concentration 1.62mg/mL
Format Affinity Purified
Quantity 0.1mL
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Primary or Secondary Primary
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot No. O43854]
Antigen EDIL3/DEL1
Monoclonal or Polyclonal Polyclonal