Please login to your online account to display your discounted pricing

anti-EDIL3/DEL1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot No. O43854]
Antigen EDIL3/DEL1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP128632

 
novus biologicals canada
NBP128632
Unlabeled 0.1mL Each for $329.00

NBP128632SS

 
novus biologicals canada
NBP128632SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot No. O43854]
Antigen EDIL3/DEL1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Gene Accession No. O43854
Gene ID (Entrez) 10085
Formulation PBS, 30% glycerol
Concentration 1.62mg/mL