missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDG2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112306

Catalog No. 01-674-106



Description

Description

Lysophosphatidic acid receptor 1 (LPA1, EDG2, VZG-1) is a seven transmembrane receptor coupled to G proteins and has high affinity to lypophosphatidic acid (LPA). LPA1 is a member of the EDG-family of receptors which includes LPA1, LPA2 and LPA3. These receptors mediate the serum borne lipid mediator LPA for a variety biological functions. The activation of LPA1 by LPA has shown to stimulate cell spreading and motility, mitogenesis, and various other nervous system activities.
Specifications

Specifications

EDG2
Polyclonal
PBS with 0.08% sodium azide
Q92633
LPA1, EDG2, VZG-1, LPA receptor 1, LPA-1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1902
Western Blot
Unconjugated
LPAR1
Liquid
LPAR1
Synthetic peptide corresponding to the C-terminal of EDG2 receptor.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
SDS
Documents