Please login to your online account to display your discounted pricing

anti-EDG2 Polyclonal, Thermo Scientific™

EDG2 Polyclonal antibody PA1-12306 from Thermo Scientific™ specifically detects EDG2 in Human samples, and it is validated for Western Blot

$723.55 - $723.55

Specifications

Antigen EDG2
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal of EDG2 receptor.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-106

 
pierce biotechnology inc
PA112306
Unlabeled 100μg Each for $723.55
Description & Specifications

Specifications

Antigen EDG2
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal of EDG2 receptor.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

EDG2 Polyclonal antibody PA1-12306 from Thermo Scientific™ specifically detects EDG2 in Human samples, and it is validated for Western Blot

Lysophosphatidic acid receptor 1 (LPA1, EDG2, VZG-1) is a seven transmembrane receptor coupled to G proteins and has high affinity to lypophosphatidic acid (LPA). LPA1 is a member of the EDG-family of receptors which includes LPA1, LPA2 and LPA3. These receptors mediate the serum borne lipid mediator LPA for a variety biological functions. The activation of LPA1 by LPA has shown to stimulate cell spreading and motility, mitogenesis, and various other nervous system activities.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the C-terminal of EDG2 receptor.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: LPA1, EDG2, VZG-1, LPA receptor 1, LPA-1
Uniprot Accession: Q92633
Gene Symbol: LPAR1
Entrez GeneID: 1902
PA1-12306 detects EDG2 in Human samples. PA1-12306 has been successfully used in Western Blot procedures. PA1-12306 The immunogen is Synthetic peptide corresponding to the C-terminal of EDG2 receptor..