Please login to your online account to display your discounted pricing

anti-EBI2/GPR183, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human EBI2 was used as the immunogen.
Antigen EBI2/GPR183
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227352SS

 
novus biologicals canada
NBP227352SS
Unlabeled 0.025mg Each for $109.00

NBP227352

 
novus biologicals canada
NBP227352
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human EBI2 was used as the immunogen.
Antigen EBI2/GPR183
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 93% homologous in mouse and rat.
Gene Accession No. P32249
Gene ID (Entrez) 1880
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL