Please login to your online account to display your discounted pricing

anti-EB3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056441

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056441

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Test Specificity In human brain, an approx. 38 kDa band is observed.
Antigen EB3
Format Purified
Conjugate Unlabeled
Gene Accession No. Q9UPY8
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg
Gene ID (Entrez) 22924
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Western Blotting
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide to a portion of the human EB3 protein.
Primary or Secondary Primary
Regulatory Status RUO