missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-EB3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide to a portion of the human EB3 protein.
Antigen EB3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056441 Novus Biologicals Canada
NB10056441
0.1mg Each for $255.80
NB10056441T Novus Biologicals Canada
NB10056441SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
EB3
Purified
RUO
In human brain, an approx. 38 kDa band is observed.
22924
0.5mg/mL
This antibody was developed against a synthetic peptide to a portion of the human EB3 protein.
Polyclonal
Primary
Western Blot
Q9UPY8
PBS containing 0.2% gelatin