Please login to your online account to display your discounted pricing

anti-EB3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056441SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056441T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9UPY8
Concentration 0.5mg/mL
Applications Western Blotting
Gene ID (Entrez) 22924
Formulation PBS containing 0.2% gelatin
Test Specificity In human brain, an approx. 38 kDa band is observed.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen EB3
Primary or Secondary Primary
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide to a portion of the human EB3 protein.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Format Purified