Please login to your online account to display your discounted pricing

anti-EAAT1/GLAST-1/SLC1A3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142587SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142587SS

  • / Each
 

Description & Specifications

Specifications

Antigen EAAT1/GLAST-1/SLC1A3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Gene Accession No. P43003
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P43003]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6507
Test Specificity Human, Mouse, Rat. Predicted to react with bovine based on 100% sequence homology.
Target Species Human
Target Species Murine
Target Species Rat