Please login to your online account to display your discounted pricing

anti-EAAT1/GLAST-1/SLC1A3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142587

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142587

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. P43003
Primary or Secondary Primary
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 6507
Format Affinity Purified
Formulation PBS
Test Specificity Human, Mouse, Rat. Predicted to react with bovine based on 100% sequence homology.
Concentration 1.0mg/mL
Antigen EAAT1/GLAST-1/SLC1A3
Quantity 0.1mL
Immunogen A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P43003]
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat