Please login to your online account to display your discounted pricing

anti-EAAT1/GLAST-1/SLC1A3, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P43003]
Antigen EAAT1/GLAST-1/SLC1A3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142587SS

 
novus biologicals canada
NBP142587SS
Unlabeled 0.025mL Each for $109.00

NBP142587

 
novus biologicals canada
NBP142587
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P43003]
Antigen EAAT1/GLAST-1/SLC1A3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human, Mouse, Rat. Predicted to react with bovine based on 100% sequence homology.
Gene Accession No. P43003
Gene ID (Entrez) 6507
Formulation PBS
Concentration 1.0mg/mL