Please login to your online account to display your discounted pricing

anti-E-Tag Epitope Tag, Polyclonal, Novus Biologicals™

$274.59 - $274.59

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen E-Tag Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600527

 
novus biologicals canada
NB600527
Unlabeled 0.1mg Each for $274.59
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen E-Tag Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Formulation phosphate Buffered Saline (PBS)
Concentration 1.0mg/mL