Please login to your online account to display your discounted pricing

anti-E-Tag Epitope Tag, Polyclonal, Novus Biologicals™

$990.46 - $990.46

Specifications

Host Species Goat
Isotype IgG
Immunogen Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH.
Antigen E-Tag Epitope Tag
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600518

 
novus biologicals canada
NB600518
Unlabeled 1mg Each for $990.46
Description & Specifications

Specifications

Host Species Goat
Isotype IgG
Immunogen Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH.
Antigen E-Tag Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation phosphate Buffered Saline (PBS)
Concentration 1.0mg/mL