Please login to your online account to display your discounted pricing

anti-E-Tag Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6005271MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6005271MG

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate Buffered Saline (PBS)
Cross Reactivity Non-species specific
Quantity 1mg
Concentration 1.0mg/mL
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen Rabbits were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Antigen E-Tag Epitope Tag