Please login to your online account to display your discounted pricing

anti-E-Tag Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB60051801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB60051801

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate Buffered Saline (PBS)
Primary or Secondary Primary
Host Species Goat
Concentration 1.0mg/mL
Format Affinity Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Isotype IgG
Conjugate Unlabeled
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Cross Reactivity Non-species specific
Immunogen Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH.
Antigen E-Tag Epitope Tag