Please login to your online account to display your discounted pricing

anti-E-Tag Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600518

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600518

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Concentration 1.0mg/mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation phosphate Buffered Saline (PBS)
Host Species Goat
Antigen E-Tag Epitope Tag
Format Affinity Purified
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Quantity 1mg
Immunogen Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH.
Cross Reactivity Non-species specific