Please login to your online account to display your discounted pricing

anti-DYRK3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11094876

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11094876

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8444
Test Specificity Human and monkey. Does not react with mouse and rat. Immunogen sequence has 94% homology to bovine.
Regulatory Status RUO
Format Affinity Purified
Conjugate Unlabeled
Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. O43781
Antigen DYRK3
Immunogen Synthetic peptide made to an internal portion of human DYRK3 (within residues 500-600). [Swiss-Prot No. O43781]
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL